1-(6-Chloro-3-nitropyridin-2-yl)azepane ≥95%
Supplier: Matrix Scientific
058215-500MG
058215-1G
101914-252EA
278.46
USD
101914-252
101914-254
1-(6-Chloro-3-nitropyridin-2-yl)azepane ≥95%
1-(6-Chloro-3-nitropyridin-2-yl)azepane
Hazard: Irritant
Formula:
C₁₁H₁₄ClN₃O₂ |
MDL Number:
MFCD11206177 CAS Number: 1094400-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...