Ethyl-4-hydroxy-2-oxo-1-phenyl-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

058127-1G
101914-044EA 903.08 USD
101914-044
Ethyl-4-hydroxy-2-oxo-1-phenyl-1,2-dihydro-1,8-naphthyridine-3-carboxylate ≥95%
Ethyl-4-hydroxy-2-oxo-1-phenyl-1,2-dihydro-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₄N₂O₄
MDL Number: MFCD17011902
CAS Number: 138305-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR