5,15-Diphenylporphyrin ≥95%

Supplier: Matrix Scientific

Synonyms: 5,15-Diphenyl-21H,23H-porphine

058084-1G
101914-004EA 1495.38 USD
101914-004
5,15-Diphenylporphyrin ≥95%
5,15-Diphenylporphyrin

Hazard: Irritant

Formula: C₃₂H₂₂N₄
MW: 462.56 g/mol
MDL Number: MFCD08276335
CAS Number: 22112-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR