N-(2-Hydroxyethyl)oleamide ≥95%

Supplier: Matrix Scientific

057785-1G
101913-614EA 53.85 USD
101913-614
N-(2-Hydroxyethyl)oleamide ≥95%
N-(2-Hydroxyethyl)oleamide

Hazard: Irritant

Formula: C₂₀H₃₉NO₂
MDL Number: MFCD00045972
CAS Number: 111-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR