5-Amino-2-chlorobenzoic acid ≥95%

Supplier: Matrix Scientific
Warning

057675-100G
101913-526EA 70.77 USD
101913-526
5-Amino-2-chlorobenzoic acid ≥95%
5-Amino-2-chlorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 188 °C
Density: 1.519 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007845
CAS Number: 89-54-3
EINECS: 201-916-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR