2-Chloro-5-(trifluoromethyl)-4-iodopyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-iodo-5-(trifluoromethyl)pyridine

057370-5G
101913-062EA 64.62 USD
101913-062
2-Chloro-5-(trifluoromethyl)-4-iodopyridine ≥95%
2-Chloro-5-(trifluoromethyl)-4-iodopyridine

Hazard: Irritant

Formula: C₆H₂ClF₃IN
MDL Number: MFCD01862099
CAS Number: 505084-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR