2,5-Dimethoxybenzyl thiocyanate ≥98%

Supplier: Matrix Scientific

057027-10G 057027-50G
101912-096EA 156.92 USD
101912-096 101912-098
2,5-Dimethoxybenzyl thiocyanate ≥98%
2,5-Dimethoxybenzyl thiocyanate

Hazard: Irritant

Formula: C₁₀H₁₁NO₂S
MW: 209.27 g/mol
MDL Number: MFCD09025688
CAS Number: 1252046-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR