2,5-Diiodobenzhydrazide

Supplier: Matrix Scientific

057020-1G 057020-5G
101912-068EA 87.69 USD
101912-068 101912-070
2,5-Diiodobenzhydrazide
2,5-Diiodobenzhydrazide

Hazard: Irritant

Formula: C₇H₆I₂N₂O
Melting Pt: 215…217 °C
MDL Number: MFCD03425684
CAS Number: 1023444-89-4

Order Now

Specification Test Results

Melting point 215-217°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR