4-Benzyloxybenzyl alcohol for synthesis
Supplier: Matrix Scientific
Synonyms:
(4-(Benzyloxy)phenyl)methanol
056895-10G
056895-50G
056895-250G
101911-628EA
83.08
USD
101911-628
101911-630
101911-632
4-Benzyloxybenzyl alcohol for synthesis
4-Benzyloxybenzyl alcohol
Hazard code(s): Irritant
Formula:
C₁₄H₁₄O₂ Melting Pt: 86…87 °C |
MDL Number:
MFCD00004654 CAS Number: 836-43-1 EINECS: 212-649-9 |
Specification Test Results
Melting point | 86-87°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...