4-Benzyloxybenzyl alcohol for synthesis

Supplier: Matrix Scientific

Synonyms: (4-(Benzyloxy)phenyl)methanol

056895-10G 056895-50G 056895-250G
101911-628EA 83.08 USD
101911-628 101911-630 101911-632
4-Benzyloxybenzyl alcohol for synthesis
4-Benzyloxybenzyl alcohol

Hazard code(s): Irritant

Formula: C₁₄H₁₄O₂
Melting Pt: 86…87 °C
MDL Number: MFCD00004654
CAS Number: 836-43-1
EINECS: 212-649-9

Order Now

Specification Test Results

Melting point 86-87°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR