Benzyl-4-hydroxybenzoate
Supplier: Matrix Scientific
|
Warning
|
056894-25G
056894-100G
101911-624EA
116.92
USD
101911-624
101911-626
Benzyl-4-hydroxybenzoate
Benzyl 4-hydroxybenzoate
Hazard code(s): Irritant
Formula:
C₁₄H₁₂O₃ Melting Pt: 110…112 °C |
MDL Number:
MFCD00016471 CAS Number: 94-18-8 EINECS: 202-311-9 |
Specification Test Results
Melting point | 110-112°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...