1-Piperidinethiocarboxamide ≥98%

Supplier: Matrix Scientific

Synonyms: Piperidine-1-carbothioamide

056817-5G
101911-462EA 253.85 USD
101911-462
1-Piperidinethiocarboxamide ≥98%
1-Piperidinethiocarboxamide

Hazard: Irritant

Formula: C₆H₁₂N₂S
Melting Pt: 127…129 °C
MDL Number: MFCD00041230
CAS Number: 14294-09-8

Order Now

Specification Test Results

Melting point 127-129°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR