Methyl-3-amino-4-(1H-pyrrol-1-yl)benzoate ≥95%

Supplier: Matrix Scientific

056496-500MG 056496-1G
101910-738EA 375.38 USD
101910-738 101910-740
Methyl-3-amino-4-(1H-pyrrol-1-yl)benzoate ≥95%
Methyl-3-amino-4-(1H-pyrrol-1-yl)benzoate

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
Melting Pt: 88…90 °C
MDL Number: MFCD16890131
CAS Number: 1242267-86-2

Order Now

Specification Test Results

Melting point 88-90°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR