tert-Butyl-3-(hydroxymethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(hydroxymethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

056456-500MG 056456-1G
101910-570EA 556.92 USD
101910-570 101910-572
tert-Butyl-3-(hydroxymethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate ≥95%
tert-Butyl-3-(hydroxymethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₃
Melting Pt: 146…148 °C
MDL Number: MFCD11054204
CAS Number: 144657-67-0

Order Now

Specification Test Results

Melting point 146-148°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR