Methyl-4-amino-5-bromo-2-hydroxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-amino-5-bromo-2-hydroxybenzoate

056420-500MG 056420-1G
101910-442EA 556.92 USD
101910-442 101910-444
Methyl-4-amino-5-bromo-2-hydroxybenzoate ≥95%
Methyl-4-amino-5-bromo-2-hydroxybenzoate

Hazard: Irritant

Formula: C₈H₈BrNO₃
MW: 246.07 g/mol
Melting Pt: 139…141 °C
MDL Number: MFCD16710276
CAS Number: 286377-20-6

Order Now

Specification Test Results

Melting point 139-141°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR