1-(6-Iodo-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazin-1-yl)-2,2-dimethylpropan-1-one

Supplier: Matrix Scientific

055667-100MG
101909-890EA 364.62 USD
101909-890
1-(6-Iodo-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazin-1-yl)-2,2-dimethylpropan-1-one
1-(6-Iodo-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazin-1-yl)-2,2-dimethylpropan-1-one

Hazard: Irritant

Formula: C₁₂H₁₅IN₂O₂
MDL Number: MFCD16628268
CAS Number: 1228665-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR