6-(3-Iodopyridin-4-yloxy)furo[3,2-b]pyridine

Supplier: Matrix Scientific

055660-100MG
101909-876EA 364.62 USD
101909-876
6-(3-Iodopyridin-4-yloxy)furo[3,2-b]pyridine
6-(3-Iodopyridin-4-yloxy)furo[3,2-b]pyridine

Hazard: Irritant

Formula: C₁₂H₇IN₂O₂
MDL Number: MFCD15530317
CAS Number: 1228665-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR