tert-Butyl-4-chloro-2-(3-hydroxyprop-1-ynyl)pyridin-3-yl carbonate

Supplier: Matrix Scientific

055585-100MG
101909-726EA 364.62 USD
101909-726
tert-Butyl-4-chloro-2-(3-hydroxyprop-1-ynyl)pyridin-3-yl carbonate
tert-Butyl-4-chloro-2-(3-hydroxyprop-1-ynyl)pyridin-3-yl carbonate

Hazard: Irritant

Formula: C₁₃H₁₄ClNO₄
MDL Number: MFCD15530284
CAS Number: 1228665-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR