3-Bromo-2-fluoro-6-(pyrrolidin-1-yl)pyridine

Supplier: Matrix Scientific

055535-100MG
101909-626EA 364.62 USD
101909-626
3-Bromo-2-fluoro-6-(pyrrolidin-1-yl)pyridine
3-Bromo-2-fluoro-6-(pyrrolidin-1-yl)pyridine

Hazard: Irritant

Formula: C₉H₁₀BrFN₂
MW: 245.1 g/mol
MDL Number: MFCD15530261
CAS Number: 1228666-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR