6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoronicotinonitrile

Supplier: Matrix Scientific

055530-100MG
101909-616EA 364.62 USD
101909-616
6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoronicotinonitrile
6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoronicotinonitrile

Hazard: Irritant

Formula: C₁₇H₂₆FN₃OSi
MW: 335.5 g/mol
MDL Number: MFCD15530256
CAS Number: 1228666-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR