3-(Trifluoromethyl)cyclohexanol cis- and trans-mixture ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)cyclohexanol, cis/trans

054924-1G 054924-5G
101909-082EA 121.54 USD
101909-082 101909-084
3-(Trifluoromethyl)cyclohexanol cis- and trans-mixture ≥97%
3-(Trifluoromethyl)cyclohexanol

Hazard: Irritant

Formula: C₇H₁₁F₃O
MDL Number: MFCD00102143
CAS Number: 454-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR