4-(1-Naphthyl)piperidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Naphthalen-1-yl)piperidine hydrochloride

054816-500MG 054816-1G
101908-648EA 921.54 USD
101908-648 101908-650
4-(1-Naphthyl)piperidine hydrochloride ≥95%
4-(1-Naphthyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₈ClN
MW: 247.77 g/mol
MDL Number: MFCD02179088
CAS Number: 314083-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR