1-(2-Aminoethyl)-4-phenylpyrazole ≥95%

Supplier: Matrix Scientific

054671-500MG 054671-1G
101908-070EA 556.92 USD
101908-070 101908-072
1-(2-Aminoethyl)-4-phenylpyrazole ≥95%
1-(2-Aminoethyl)-4-phenylpyrazole

Hazard: Irritant

Formula: C₁₁H₁₃N₃
MDL Number: MFCD16872049
CAS Number: 1211530-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR