2-Chloro-5-(trifluoromethyl)pyridin-3-amine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Amino-2-chloro-5-(trifluoromethyl)pyridine

054655-1G 054655-5G
101908-008EA 53.85 USD
101908-008 101908-010
2-Chloro-5-(trifluoromethyl)pyridin-3-amine ≥98%
2-Chloro-5-(trifluoromethyl)pyridin-3-amine

Hazard: Irritant

Formula: C₆H₄ClF₃N₂
MDL Number: MFCD07375383
CAS Number: 72587-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR