1-Thia-4,8-diazaspiro[4.5]decan-3-one hydrochloride ≥95%

Supplier: Matrix Scientific

054319-500MG
101907-052EA 556.92 USD
101907-052
1-Thia-4,8-diazaspiro[4.5]decan-3-one hydrochloride ≥95%
1-Thia-4,8-diazaspiro[4.5]decan-3-one hydrochloride

Hazard: Irritant

Formula: C₇H₁₃ClN₂OS
MW: 208.71 g/mol
MDL Number: MFCD14581672
CAS Number: 1203898-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR