2-Cyclopropyl-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol hydrochloride

Supplier: Matrix Scientific

053879-500MG 053879-2.5G
101905-328EA 364.62 USD
101905-328 101905-330
2-Cyclopropyl-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol hydrochloride
2-Cyclopropyl-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidin-4-ol hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClN₃O
MDL Number: MFCD13562958
CAS Number: 1220038-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR