N-(4-Fluorophenyl)-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053506-500MG 053506-2.5G
101903-910EA 364.62 USD
101903-910 101903-912
N-(4-Fluorophenyl)-4-piperidinecarboxamide hydrochloride
N-(4-Fluorophenyl)-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClFN₂O
MDL Number: MFCD13562831
CAS Number: 1220038-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR