N-(3-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053425-500MG 053425-2.5G
101903-586EA 364.62 USD
101903-586 101903-588
N-(3-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride
N-(3-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O
MDL Number: MFCD13562776
CAS Number: 1246172-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR