2-[(6-Chloro-2-pyrazinyl)amino]-1-ethanol
Supplier: Matrix Scientific
052866-500MG
052866-2.5G
101901-352EA
364.62
USD
101901-352
101901-354
2-[(6-Chloro-2-pyrazinyl)amino]-1-ethanol
2-[(6-Chloro-2-pyrazinyl)amino]-1-ethanol
Hazard: Irritant
Formula:
C₆H₈ClN₃O |
MDL Number:
MFCD13562417 CAS Number: 1147998-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...