4-(Allylamino)-3-nitrobenzoic acid

Supplier: Matrix Scientific

052812-500MG 052812-2.5G
101901-136EA 364.62 USD
101901-136 101901-138
4-(Allylamino)-3-nitrobenzoic acid
4-(Allylamino)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₄
MDL Number: MFCD11131943
CAS Number: 160219-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR