3,4-Dihydro-2(1H)-isoquinolinyl(4-piperidinyl)methanone hydrochloride

Supplier: Matrix Scientific

052498-500MG 052498-2.5G
101899-878EA 364.62 USD
101899-878 101899-880
3,4-Dihydro-2(1H)-isoquinolinyl(4-piperidinyl)methanone hydrochloride
3,4-Dihydro-2(1H)-isoquinolinyl(4-piperidinyl)methanone hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₁ClN₂O
MDL Number: MFCD09863335
CAS Number: 73415-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR