2-Amino-1-(4-benzyl-1-piperazinyl)-3-phenyl-1-propanone hydrochloride

Supplier: Matrix Scientific

052374-500MG 052374-2.5G
101899-382EA 364.62 USD
101899-382 101899-384
2-Amino-1-(4-benzyl-1-piperazinyl)-3-phenyl-1-propanone hydrochloride
2-Amino-1-(4-benzyl-1-piperazinyl)-3-phenyl-1-propanone hydrochloride

Hazard: Irritant

Formula: C₂₀H₂₆ClN₃O
MDL Number: MFCD13562133
CAS Number: 1236266-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR