2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-1-propanone hydrochloride

Supplier: Matrix Scientific

052242-500MG 052242-2.5G
101898-854EA 166.15 USD
101898-854 101898-856
2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-1-propanone hydrochloride
2-Amino-1-[3,4-dihydro-1(2H)-quinolinyl]-1-propanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MW: 240.74 g/mol
MDL Number: MFCD13562015
CAS Number: 1236260-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR