4-(4-Benzyl-1-piperazinyl)-6-chloro-1,3,5-triazin-2-ylamine

Supplier: Matrix Scientific

052087-500MG 052087-2.5G
101898-238EA 186.15 USD
101898-238 101898-240
4-(4-Benzyl-1-piperazinyl)-6-chloro-1,3,5-triazin-2-ylamine
4-(4-Benzyl-1-piperazinyl)-6-chloro-1,3,5-triazin-2-ylamine

Hazard: Irritant

Formula: C₁₄H₁₇ClN₆
MDL Number: MFCD13561895
CAS Number: 1220029-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR