[6-(1-Piperidinyl)-3-pyridinyl]methanamine

Supplier: Matrix Scientific

052049-500MG 052049-2.5G
101898-086EA 461.54 USD
101898-086 101898-088
[6-(1-Piperidinyl)-3-pyridinyl]methanamine
[6-(1-Piperidinyl)-3-pyridinyl]methanamine

Hazard: Irritant

Formula: C₁₁H₁₇N₃
MDL Number: MFCD08235199
CAS Number: 914637-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR