(4-Benzyl-1-piperazinyl)(2-chloro-3-pyridinyl)methanone

Supplier: Matrix Scientific

052030-500MG 052030-2.5G
101898-010EA 364.62 USD
101898-010 101898-012
(4-Benzyl-1-piperazinyl)(2-chloro-3-pyridinyl)methanone
(4-Benzyl-1-piperazinyl)(2-chloro-3-pyridinyl)methanone

Hazard: Irritant

Formula: C₁₇H₁₈ClN₃O
MDL Number: MFCD03377660
CAS Number: 544428-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR