2-[4-(3-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol

Supplier: Matrix Scientific

052013-500MG 052013-2.5G
101897-942EA 364.62 USD
101897-942 101897-944
2-[4-(3-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol
2-[4-(3-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₈N₄O
MDL Number: MFCD08692318
CAS Number: 1082137-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR