2-(1-Pyrrolidinyl)nicotinic acid

Supplier: Matrix Scientific

051967-500MG 051967-2.5G
101897-758EA 364.62 USD
101897-758 101897-760
2-(1-Pyrrolidinyl)nicotinic acid
2-(1-Pyrrolidinyl)nicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MDL Number: MFCD05865128
CAS Number: 690632-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR