1-(6-Chloro-4-pyrimidinyl)azepane

Supplier: Matrix Scientific

051844-500MG 051844-2.5G
101897-268EA 364.62 USD
101897-268 101897-270
1-(6-Chloro-4-pyrimidinyl)azepane
1-(6-Chloro-4-pyrimidinyl)azepane

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MDL Number: MFCD13561822
CAS Number: 1220033-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR