Ethyl-3-amino-4-(diallylamino)benzoate

Supplier: Matrix Scientific

051731-500MG 051731-2.5G
101896-816EA 364.62 USD
101896-816 101896-818
Ethyl-3-amino-4-(diallylamino)benzoate
Ethyl-3-amino-4-(diallylamino)benzoate

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₂
MDL Number: MFCD13561775
CAS Number: 1219967-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR