N-Benzyl-N-methyl-2-(3-piperidinyl)-1-ethanamine dihydrochloride

Supplier: Matrix Scientific

051600-500MG
101896-292EA 364.62 USD
101896-292
N-Benzyl-N-methyl-2-(3-piperidinyl)-1-ethanamine dihydrochloride
N-Benzyl-N-methyl-2-(3-piperidinyl)-1-ethanamine dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₆Cl₂N₂
MDL Number: MFCD13561654
CAS Number: 1220035-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR