4-(4-Piperidinyl)morpholine dihydrochloride

Supplier: Matrix Scientific

Synonyms: 4-(4-Piperidinyl)morpholine dihydrochloride

051498-500MG 051498-2.5G
101895-884EA 364.62 USD
101895-884 101895-886
4-(4-Piperidinyl)morpholine dihydrochloride
4-(Piperidin-4-yl)morpholine dihydrochloride

Hazard: Irritant

Formula: C₉H₂₀Cl₂N₂O
MDL Number: MFCD09033209
CAS Number: 550370-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR