2-[Methyl-(3-piperidinylmethyl)amino]-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051412-500MG 051412-2.5G
101895-540EA 364.62 USD
101895-540 101895-542
2-[Methyl-(3-piperidinylmethyl)amino]-1-ethanol dihydrochloride
2-[Methyl-(3-piperidinylmethyl)amino]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₉H₂₂Cl₂N₂O
MDL Number: MFCD13561486
CAS Number: 1219957-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR