2-[2-Amino(ethyl)-4-fluoroanilino]-1-ethanol

Supplier: Matrix Scientific

051204-500MG 051204-2.5G
101894-708EA 364.62 USD
101894-708 101894-710
2-[2-Amino(ethyl)-4-fluoroanilino]-1-ethanol
2-[2-Amino(ethyl)-4-fluoroanilino]-1-ethanol

Hazard: Irritant

Formula: C₁₀H₁₅FN₂O
MDL Number: MFCD12772397
CAS Number: 1179629-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR