Methyl (2S,4S)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

051039-500MG 051039-2.5G
101894-048EA 364.62 USD
101894-048 101894-050
Methyl (2S,4S)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-(3,4-dimethylphenoxy)-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD13561409
CAS Number: 1354487-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR