2-Iodophenyl-4-piperidinyl ether hydrochloride

Supplier: Matrix Scientific

050804-500MG 050804-2.5G
101893-112EA 364.62 USD
101893-112 101893-114
2-Iodophenyl-4-piperidinyl ether hydrochloride
2-Iodophenyl-4-piperidinyl ether hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClINO
MDL Number: MFCD13561184
CAS Number: 1220019-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR