2-[2-(3-Nitrophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050191-500MG 050191-2.5G
101890-664EA 364.62 USD
101890-664 101890-666
2-[2-(3-Nitrophenoxy)ethyl]piperidine hydrochloride
2-[2-(3-Nitrophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₃
MDL Number: MFCD13560592
CAS Number: 1220029-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR