2-[2-(4-Methyl-2-nitrophenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

050187-500MG 050187-2.5G
101890-648EA 364.62 USD
101890-648 101890-650
2-[2-(4-Methyl-2-nitrophenoxy)ethyl]piperidine hydrochloride
2-[2-(4-Methyl-2-nitrophenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O₃
MDL Number: MFCD13560588
CAS Number: 1219982-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR