2-[2-(Allyloxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049826-500MG 049826-2.5G
101889-204EA 364.62 USD
101889-204 101889-206
2-[2-(Allyloxy)ethyl]piperidine hydrochloride
2-[2-(Allyloxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO
MW: 205.73 g/mol
MDL Number: MFCD13560228
CAS Number: 1219949-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR