4-[2-(4-Chloro-2-ethylphenoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049708-500MG 049708-2.5G
101888-732EA 364.62 USD
101888-732 101888-734
4-[2-(4-Chloro-2-ethylphenoxy)ethyl]piperidine hydrochloride
4-[2-(4-Chloro-2-ethylphenoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560110
CAS Number: 1219964-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR